Learn More
Hydrazine sulfate, 99+%, Thermo Scientific Chemicals
$110.05 - $297.84
Chemical Identifiers
CAS | 10034-93-2 |
---|---|
Molecular Formula | H4N2·H2SO4 |
Molecular Weight (g/mol) | 130.12 |
MDL Number | MFCD00044873 |
InChI Key | ZGCHATBSUIJLRL-UHFFFAOYSA-N |
Synonym | hydrazine sulfate, hydrazine monosulfate, hydrazine, sulfate, hydrazine sulphate, hydrazinium sulfate, hydrazonium sulfate, hydrazine sulfate 1:1, segidrin, sehydrin, diamine sulfate |
PubChem CID | 24842 |
IUPAC Name | hydrazine;sulfuric acid |
SMILES | NN.OS(=O)(=O)O |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC201861000
|
Thermo Scientific Chemicals
201861000 |
100 g | Plastic bottle |
Each for $110.05
|
|
||||
AC201865000
|
Thermo Scientific Chemicals
201865000 |
500 g | Plastic bottle |
Each for $297.84
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Reduction reagentChemical Identifiers
10034-93-2 | |
130.12 | |
ZGCHATBSUIJLRL-UHFFFAOYSA-N | |
24842 | |
NN.OS(=O)(=O)O |
H4N2·H2SO4 | |
MFCD00044873 | |
hydrazine sulfate, hydrazine monosulfate, hydrazine, sulfate, hydrazine sulphate, hydrazinium sulfate, hydrazonium sulfate, hydrazine sulfate 1:1, segidrin, sehydrin, diamine sulfate | |
hydrazine;sulfuric acid |
Specifications
10034-93-2 | |
White | |
Plastic bottle | |
H2NNH2·H2SO4 | |
100 g | |
hydrazine sulfate, hydrazine monosulfate, hydrazine, sulfate, hydrazine sulphate, hydrazinium sulfate, hydrazonium sulfate, hydrazine sulfate 1:1, segidrin, sehydrin, diamine sulfate | |
ZGCHATBSUIJLRL-UHFFFAOYSA-N | |
hydrazine;sulfuric acid | |
24842 | |
99+% | |
Hydrazine sulfate, p.a. |
254.0°C | |
99+% | |
H4N2·H2SO4 | |
MFCD00044873 | |
15, 4810 | |
Solubility in water: 30g/L (20°C). Other solubilities: insoluble in alcohol | |
NN.OS(=O)(=O)O | |
130.12 | |
130.12 | |
Crystalline Powder |
Safety and Handling
GHS H Statement
May cause cancer.
Toxic in contact with skin.
Toxic if inhaled.
Toxic if swallowed.
May cause an allergic skin reaction.
Very toxic to aquatic life with long lasting effects.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fre
GHS Signal Word: Danger
EINECSNumber : 233-110-4
RTECSNumber : MV9625000
TSCA : TSCA
RUO – Research Use Only