Please login to your online account to display your discounted pricing

Alfa Aesar™ Hydrazine, standard solution, Specpure™, N2H4 100μg/mL

Chemical Identifiers

Chemical Name or Material Hydrazine, standard solution
Name Note N2H4 100μg/mL
Purity Grade Specpure™
Formula Weight 50.06
MDL Number MFCD00011417
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA42561AO

  SDS
alfa aesar™
42561AO
450mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 50.06
Quantity 450mL
Physical Form Liquid
Chemical Name or Material Hydrazine, standard solution
Name Note N2H4 100μg/mL
Odor Ammonia-like
MDL Number MFCD00011417
Purity Grade Specpure™
TSCA TSCA