Please login to your online account to display your discounted pricing

Hydrazine Standard, Certified, 1000ppm ±10ppm (1mL = 1mg N2H4), LabChem™

$52.83 - $52.83

Chemical Identifiers

Chemical Name or Material Hydrazine Standard
CAS 7732-18-5
CAS Max % 99.17
CAS 7647-01-0
CAS Max % 0.5
CAS 5341-61-7
CAS Max % 0.33
Name Note 1000ppm (1mL = 1mg N2H4)
Purity Grade Certified Grade
Linear Formula N2H4·2HCl
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

LC149301

 
lab chem
LC149301
Poly Bottle 500mL Each for $52.83
Description & Specifications

Specifications

Purity Grade Certified Grade
Formula Weight 104.97
Health Hazard 3 GHS P Statement
Obtain special instructions before use.
Do not handle until all safety precautions have been read and understood.
Do not breathe mist, vapors, spray.
Wear protective gloves, eye protection.
Contaminated work clothing should not be allowed out of the workplace.
If exposed or concerned: Get medical advice/attention.
If on skin (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
If skin irritation or rash occurs: Get medical advice/attention.
Store locked up.
Dispose of contents/container to comply with local, state and federal regulations.
Identification Passes Test
CAS 5341-61-7
Health Hazard 2 GHS H Statement
May cause an allergic skin reaction.
May cause cancer.
Harmful to the environment with long lasting effects.
Solubility Soluble in water
Physical Form Liquid
Health Hazard 1 Danger
Traceability to NIST Traceable to NIST
Name Note 1000ppm (1mL = 1mg N2H4)
Density 1g/mL
CAS Max % 0.33
CAS Max % 99.17
Concentration 1000ppm ±10ppm (1mL = 1mg N2H4)
Linear Formula N2H4·2HCl
Quantity 500mL
Color Clear, colorless
Recommended Storage Room Temperature
UN Number UN1789
CAS 7732-18-5
Chemical Name or Material Hydrazine Standard
CAS 7647-01-0
CAS Max % 0.5
Packaging Poly Bottle
Safety and Handling

Danger

GHS P Statement
Obtain special instructions before use.
Do not handle until all safety precautions have been read and understood.
Do not breathe mist, vapors, spray.
Wear protective gloves, eye protection.
Contaminated work clothing should not be allowed out of the workplace.
If exposed or concerned: Get medical advice/attention.
If on skin (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
If skin irritation or rash occurs: Get medical advice/attention.
Store locked up.
Dispose of contents/container to comply with local, state and federal regulations.

GHS H Statement
May cause an allergic skin reaction.
May cause cancer.
Harmful to the environment with long lasting effects.

Room Temperature