Learn More
Hydrazine monohydrochloride, 98+%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 201992500
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Reduction reagentSpecifications
Hydrazine monohydrochloride | |
2644-70-4 | |
Powder | |
98% min. (Argentometry) | |
H2NNH2·HCl | |
hydrazine hydrochloride, hydrazine monohydrochloride, hydrazine, monohydrochloride, hydrazine,chloride, diamine hydrochloride, hydrazine.hydrochloride, hydrazine,hydrochloride, hydrazine mono hcl, nh2nh2hcl, pubchem17422 | |
BIVUUOPIAYRCAP-UHFFFAOYSA-N | |
hydrazine;hydrochloride | |
115031 | |
≥98% | |
Glass bottle |
89°C to 93°C | |
White | |
250 g | |
H4N2·HCl | |
MFCD00044368 | |
Solubility in water: 370g/L (20°C). Other solubilities: slightly soluble in alcohol,insoluble in most organic solvents,soluble in liquid ammoniak | |
NN.Cl | |
68.51 | |
68.51 | |
Authentic |
Chemical Identifiers
2644-70-4 | |
68.51 | |
BIVUUOPIAYRCAP-UHFFFAOYSA-N | |
115031 | |
NN.Cl |
H4N2·HCl | |
MFCD00044368 | |
hydrazine hydrochloride, hydrazine monohydrochloride, hydrazine, monohydrochloride, hydrazine,chloride, diamine hydrochloride, hydrazine.hydrochloride, hydrazine,hydrochloride, hydrazine mono hcl, nh2nh2hcl, pubchem17422 | |
hydrazine;hydrochloride |
Safety and Handling
GHS H Statement
May cause cancer.
Toxic if inhaled.
Toxic if swallowed.
May cause an allergic skin reaction.
Very toxic to aquatic life with long lasting effects.
Toxic in contact with skin.
GHS P Statement
Avoid release to the environment.
Wear protective gloves/protective clothing/eye protection/face protection.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Gently wash with plenty of soap and water.
GHS Signal Word: Danger
EINECSNumber : 220-154-4
RUO – Research Use Only