Please login to your online account to display your discounted pricing

Hydrazine monohydrochloride, 98+%, ACROS Organics™


Quantity 1kg
Packaging Glass bottle

Manufacturer: thermo fisher scientific  201990010

 View more versions of this product

Catalog No. AC201990010

  • / Each
 

Description & Specifications

Chemical Identifiers

Physical Form Powder
Assay ≥98.0% (Argentometry)
Formula Weight 68.51
Quantity 1kg
Color White to off-white
Melting Point 89° to 93°C
Chemical Name or Material Hydrazine monohydrochloride
Infrared Spectrum Authentic
Molecular Formula H4N2·HCl
CAS 2644-70-4
Assay Percent Range 98+%
MDL Number MFCD00044368
Health Hazard 1 GHS Signal Word: Danger
Health Hazard 2 GHS H Statement:
May cause cancer.
Toxic if inhaled.
Toxic if swallowed.
May cause an allergic skin reaction.
Very toxic to aquatic life with long lasting effects.
Toxic in contact with skin.
Packaging Glass bottle
Linear Formula H2NNH2·HCl
Health Hazard 3 GHS P Statement:
Avoid release to the environment.
Wear protective gloves/protective clothing/eye protection/face protection.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Gently wash with plenty of soap and water.
Obtain special instructions before use.
IF exposed or concerned: Get medical advice/attention.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Specifications

Chemical Name or Material Hydrazine monohydrochloride
Assay ≥98.0% (Argentometry)
Assay Percent Range 98+%
Molecular Formula H4N2·HCl
Linear Formula H2NNH2·HCl
Formula Weight 68.51
MDL Number MFCD00044368
Accessories

GHS Signal Word: Danger

GHS P Statement:
Avoid release to the environment.
Wear protective gloves/protective clothing/eye protection/face protection.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Gently wash with plenty of soap and water.
Obtain special instructions before use.
IF exposed or concerned: Get medical advice/attention.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
May cause cancer.
Toxic if inhaled.
Toxic if swallowed.
May cause an allergic skin reaction.
Very toxic to aquatic life with long lasting effects.
Toxic in contact with skin.

SDS & Safety Documents