Please login to your online account to display your discounted pricing

Alfa Aesar™ Hydrazine monohydrobromide, 98%

Chemical Identifiers

Chemical Name or Material Hydrazine monohydrobromide
CAS 13775-80-9
Molecular Formula H2NNH2·HBr
Formula Weight 112.96g/mol
MDL Number MFCD00065394
UN Number UN2928
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1946218

 
alfa aesar™
A19462-18
50g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAA1946230

 
alfa aesar™
A19462-30
250g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Boiling Point ≈190°C (dec.)
Melting Point 92° to 93°C
Quantity 50g
Chemical Name or Material Hydrazine monohydrobromide
CAS 13775-80-9
Assay 98%
Molecular Formula H2NNH2·HBr
Formula Weight 112.96g/mol
MDL Number MFCD00065394
UN Number UN2928
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H301-H311-H331-H350-H317
Toxic if swallowed.
Toxic in contact with skin.
Toxic if inhaled.
May cause cancer.
May cause an allergic skin reaction.
Health Hazard 3 GHS P Statement
P261-P280-P301+P310-P361-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: TOXIC SOLIDS, CORROSIVE, ORGANIC, N.O.S.
TSCA TSCA
Safety and Handling

Danger

GHS P Statement
P261-P280-P301+P310-P361-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H301-H311-H331-H350-H317
Toxic if swallowed.
Toxic in contact with skin.
Toxic if inhaled.
May cause cancer.
May cause an allergic skin reaction.