Please login to your online account to display your discounted pricing

Alfa Aesar™ Hydrazine monohydrate, 99+%

Chemical Identifiers

Chemical Name or Material Hydrazine monohydrate
CAS 7803-57-8
Molecular Formula H4N2·H2O
Linear Formula H2NNH2·H2O
Formula Weight 50.06 (32.05 Anhydrous)
MDL Number MFCD00149931
UN Number UN2030
Merck Index 14,4771
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA1665157

  SDS
alfa aesar™
1665157
4 x 500g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA16651A3

  SDS
alfa aesar™
16651A3
2kg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 50.06 (32.05 Anhydrous)
Assay Percent Range >99%
Molecular Formula H4N2·H2O
Health Hazard 3 GHS P Statement
P210-P301+P310-P303+P361+P353-P304+P340-P305+P351+P338-P320-P330-P361-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Specific treatment is urgent (see ... on this label).
Rinse mouth.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Assay ≥99%
Health Hazard 1 Danger
CAS 7803-57-8
Boiling Point 120° to 121°C
Density 1.027g/mL
Refractive Index 1.4310
Linear Formula H2NNH2·H2O
RTECS Number MV4590000
TSCA TSCA
Chemical Name or Material Hydrazine monohydrate
Flash Point 75° (167°F)
MDL Number MFCD00149931
Merck Index 14,4771
Odor Amine-like
Health Hazard 2 GHS H Statement
H301-H310-H330-H350-H314-H318-H227-H317 Toxic if swallowed.
Fatal in contact with skin.
Fatal if inhaled.
May cause cancer.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Combustible liquid.
May cause an allergic skin reaction.
DOT Information Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: HYDRAZINE AQUEOUS SOLUTION
Melting Point −52°C
UN Number UN2030
Quantity 4 x 500g
Color Colorless
Physical Form Liquid