missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Hydrazine monohydrate, 99+%, Alfa Aesar™

Manufacturer:  Alfa Aesar 16651A3

 View more versions of this product

Catalog No. AA16651A3



Specifications

Specifications

Hydrazine Monohydrate
120°C to 121°C
-52°C
7803-57-8
≥99%
H6N2O
H2NNH2·H2O
2030
14,4771
NN.O
50.061
CHEBI:35511
Liquid
75° (167°F)
Colorless
2kg
1.027g/mL
>99%
Amine-like
MFCD00149931
1.4310
IKDUDTNKRLTJSI-UHFFFAOYSA-N
hydrazine;hydrate
24654
50.06 (32.05 Anhydrous)

Chemical Identifiers

7803-57-8
50.061
IKDUDTNKRLTJSI-UHFFFAOYSA-N
CHEBI:35511
NN.O
H6N2O
MFCD00149931
24654
hydrazine;hydrate
Safety and Handling

Safety and Handling

GHS H Statement
H301-H310-H330-H350-H314-H318-H227-H317
Toxic if swallowed.
Fatal in contact with skin.
Fatal if inhaled.
May cause cancer.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Combustible liquid.
May cause an allergic skin reaction.

GHS P Statement
P210-P301+P310-P303+P361+P353-P304+P340-P305+P351+P338-P320-P330-P361-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Specific treatment is urgent (see ... on this label).
Rinse mouth.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: HYDRAZINE AQUEOUS SOLUTION

RTECSNumber : MV4590000

TSCA : TSCA

SDS Documents