Please login to your online account to display your discounted pricing

Hydrazine dihydrochloride, >98%, Alfa Aesar ™

For Use As Chlorine Scavenger in Hcl Gas Streams

$59.39 - $183.32

Chemical Identifiers

Chemical Name or Material Hydrazine dihydrochloride
Name Note Special handling precautions required
CAS 5341-61-7
Physical Form Crystalline or lump
Assay Percent Range >98%
Molecular Formula H2NNH2·2HCl
Formula Weight 104.97g/mol
MDL Number MFCD00064543
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA3272522

 
alfa aesar
32725-22
100g Each for $59.39

AA3272536

 
alfa aesar
32725-36
500g Each for $183.32
Description & Specifications

Specifications

Appearance White crystalline or lump
Density 1.42g/cm3
Odor Odorless
Color White
Quantity 100g
Chemical Name or Material Hydrazine dihydrochloride
Name Note Special handling precautions required
CAS 5341-61-7
Physical Form Crystalline or lump
Assay Percent Range >98%
Molecular Formula H2NNH2·2HCl
Formula Weight 104.97g/mol
MDL Number MFCD00064543
Health Hazard 1 H301-H311-H331-H317-H350-H400-H410: Toxic if swallowed. Toxic in contact with skin. Toxic if inhaled. May cause an allergic skin reaction. May cause cancer. Very toxic to aquatic life. Very toxic to aquatic life with long lasting effects.
DOT Information DOT Class: 6.1, Packing Group: III
EINECS Number 226-283-2
Safety and Handling

H301-H311-H331-H317-H350-H400-H410: Toxic if swallowed. Toxic in contact with skin. Toxic if inhaled. May cause an allergic skin reaction. May cause cancer. Very toxic to aquatic life. Very toxic to aquatic life with long lasting effects.