missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Hydrazine Acetate 98.0+%, TCI America™

$177.51 - $532.54

Chemical Identifiers

CAS 13255-48-6
Molecular Formula C2H8N2O2
Molecular Weight (g/mol) 92.10
MDL Number MFCD00013141
InChI Key YFHNDHXQDJQEEE-UHFFFAOYSA-N
PubChem CID 165591
IUPAC Name acetic acid; hydrazine
SMILES NN.CC(O)=O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
H11125G
View Documents Promotion Details
TCI America
H11125G
5 g
Each for $177.51
 
H111225G
View Documents Promotion Details
TCI America
H111225G
25 g
Each for $532.54
 
Description

Description

Specifications

Chemical Identifiers

13255-48-6
92.10
YFHNDHXQDJQEEE-UHFFFAOYSA-N
acetic acid; hydrazine
C2H8N2O2
MFCD00013141
165591
NN.CC(O)=O

Specifications

13255-48-6
White
MFCD00013141
3263
NN.CC(O)=O
92.10
92.10
Crystalline Powder
101°C
C2H8N2O2
5 g
YFHNDHXQDJQEEE-UHFFFAOYSA-N
acetic acid; hydrazine
165591
≥98.0% (T)
Hydrazine Acetate
Safety and Handling

Safety and Handling

TSCA : No

SDS
missing translation for 'documents'

missing translation for 'documents'