HYDRANAL™ - Water Standard 0.1, Standard for Karl Fischer titration (water content 0.1 mg/g = 0.01%), verified against NIST SRM 2890 & NMIJ CRM 4222, Honeywell™ Fluka™
Manufacturer: Honeywell-Hydranal 3484740ML
Specifications
HYDRANAL™-Water Standard 0.1 | |
25°C (77°F) | |
95-47-6,7732-18-5 |
0.86g/mL at 20°C | |
10 x 4mL ampoules | |
UN1307 |