HYDRANAL™ - Titrant 2 E, Reagent for volumetric two-component Karl Fischer titration (ethanol based), Honeywell™ Fluka™
Supplier:  Honeywell Inc 347236X1LUS
 View more versions of this product
Catalog No. 60046398
Description
Description
Specifications
Specifications
12°C
∼0.83 g/mL
Reagent for volumetric two-component Karl Fischer titration (ethanol based)
≤100.0000%,<5.0000%,<5.0000%
78°C
H2O
UN1170
Karl Fischer
6 x 1 L
HYDRANAL™ - Titrant 2 E
≥90.0000%,≥1.0000%,≥1.0000%
7553-56-2,68007-08-9
Glass Bottle
MFCD00011332
Ethyl alcohol