missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
HYDRANAL™-Humidity absorber, Honeywell Fluka™
Drying agent for air and gases for Karl Fischer titration (with indicator)
$147.48 - $1276.70
Specifications
Chemical Name or Material | HYDRANAL™-Humidity absorber |
---|---|
UN Number | NONH for all modes of transport |
Description
Advantages of HYDRANAL- Auxiliaries
- Broad product range of solubilizer for special sample application
- Broad product range of buffer for pH related sample application
- Drying agents for cell protection and drying of carrier gasses.
Specifications
HYDRANAL™-Humidity absorber | |
NONH for all modes of transport |