missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
HYDRANAL™ - Coulomat Oil, Reagent for coulometric KF titration in oils (anolyte solution), preferred for cells with diaphragm, Honeywell Fluka™
Anolyte for coulometric Karl Fischer titration in oils (chloroform-methanol-xylene-based), preferred for cells with diaphragm
$52.74 - $1,483.68
Specifications
Chemical Name or Material | HYDRANAL™-Coulomat Oil |
---|---|
Flash Point | 8°C (46.4°F) |
UN Number | UN1992 |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
6000651
|
Honeywell-Hydranal
34868100ML |
100mL |
Each for $52.74
|
|
6000652
|
Honeywell-Hydranal
348686X500MLUS |
500mL |
Each for $159.59
|
|
60050010
|
Honeywell-Hydranal
348686X100ML |
6 x 100mL |
Case of 6 Each for $456.50
|
|
60046491
|
Honeywell-Hydranal
348686X500MLUSDEL |
6 x 500mL |
Case of 6 Each for $1,483.68
|
|
Description
Advantages of Hydranal Coulometric Reagents
- Easy to use
- High accuracy for trace amounts of water
- Stable conditions of the titration vessel
- Broad product range
- Long shelf life (up to five years)
Specifications
HYDRANAL™-Coulomat Oil | |
8°C (46.4°F) | |
UN1992 |