HYDRANAL™ - Composite 1, Reagent for volumetric one-component Karl Fischer titration (methanol free), Honeywell™ Fluka™
Supplier:  Honeywell Inc 34827500MLUS
 View more versions of this product
Catalog No. 6002574
Description
Description
Specifications
Specifications
90°C (194°F)
HYDRANAL™-Composite 1
111-90-0,7446-09-5,288-32-4
500 mL
1.11g/mL