missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

HYDRANAL™ - Composite 1, Reagent for volumetric one-component Karl Fischer titration (methanol free), Honeywell™ Fluka™

$110.59 - $889.61

Specifications

Chemical Name or Material HYDRANAL™ - Composite 1
Density ∼1.11 g/mL
Flash Point 97°C
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
6002574
View Documents Promotion Details
Honeywell-Hydranal
34827500MLUS
500mL
Each for $110.59
 
6002573
View Documents Promotion Details
Honeywell-Hydranal
348276X1LUS
1L
Each for $172.71
 
60046477
View Documents Promotion Details
Honeywell-Hydranal
348276X500MLUS
6 x 500mL
Case of 6 Each for $889.61
 
Description

Description

Specifications

Specifications

HYDRANAL™ - Composite 1
∼1.11 g/mL
97°C
SDS
Product Certifications