Please login to your online account to display your discounted pricing

HYDRA-POINT™ Solvent, General Purpose, BAKER ANALYZED™ Reagent, J.T.Baker™

High quality chemicals for laboratory and specialized industrial use

$151.65 - $385.83

Chemical Identifiers

Chemical Name or Material HYDRA-POINT™ Solvent
Name Note General Purpose
Purity Grade BAKER ANALYZED™ Reagent
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

02003162

 
avantor performance
627602
Narrow Mouth Amber Glass Bottle 1L Each for $151.65

02003163

 
avantor performance
627668
Amber Glass Bottle 2.5L Each for $385.83
Description & Specifications

Specifications

Quantity 1L
Chemical Name or Material HYDRA-POINT™ Solvent
Name Note General Purpose
Purity Grade BAKER ANALYZED™ Reagent
Packaging Narrow Mouth Amber Glass Bottle
Flash Point 11.00°C

    For Karl Fischer titration