Please login to your online account to display your discounted pricing

HYDRA-POINT™ Solvent, General Purpose, BAKER ANALYZED™ Reagent, J.T.Baker™


High quality chemicals for laboratory and specialized industrial use

Quantity 2.5L
Packaging Amber Glass Bottle

Manufacturer: avantor performance  627668

 View more versions of this product

Catalog No. 02003163

  • / Each
 

Description & Specifications

Chemical Identifiers

Quantity 2.5L
Chemical Name or Material HYDRA-POINT™ Solvent
Name Note General Purpose
Purity Grade BAKER ANALYZED™ Reagent
Packaging Amber Glass Bottle
Flash Point 11.00°C

Specifications

Chemical Name or Material HYDRA-POINT™ Solvent
Name Note General Purpose
Purity Grade BAKER ANALYZED™ Reagent

    For Karl Fischer titration