Please login to your online account to display your discounted pricing

HYDRA-POINT™ Buffer Solution, BAKER ANALYZED™ Reagent, J.T.Baker™

High quality chemicals for laboratory and specialized industrial use

$191.59 - $465.52

Chemical Identifiers

Chemical Name or Material HYDRA-POINT™ Buffer Solution
Purity Grade BAKER ANALYZED™ Reagent
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

02003154

 
avantor performance
628202
Narrow Mouth Amber Glass Bottle 1L Each for $465.52

02003156

 
avantor performance
628201
Narrow Mouth Amber Glass Bottle 500mL Each for $191.59
Description & Specifications

Specifications

Quantity 1L
Flash Point 11.00°C
Chemical Name or Material HYDRA-POINT™ Buffer Solution
Purity Grade BAKER ANALYZED™ Reagent
Packaging Narrow Mouth Amber Glass Bottle

    For Karl Fischer titration