Please login to your online account to display your discounted pricing

Hydantoin, 99%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material Hydantoin
Name Note 99%
CAS 461-72-3
Assay 98.5 to 101.5%
Assay Percent Range 99%
Molecular Formula C3H4N2O2
Formula Weight 100.08
MDL Number MFCD00005259
Synonym 2, 4-Imidazolinedione
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC168701000

  SDS
fisher scientific acros
168701000
Plastic bottle 100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC168705000

  SDS
thermo fisher scientific
168705000
Plastic bottle 500g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Physical Form Fine Powder
Formula Weight 100.08
Quantity 100g
Color White to light yellow
Assay 98.5 to 101.5%
TSCA TSCA
RTECS Number MT8210000
EINECS Number 207-313-3
Melting Point 220° to 223°C
Beilstein 24, 242
Merck Index 15, 4799
Synonym 2, 4-Imidazolinedione
Infrared Spectrum Authentic
Packaging Plastic bottle
Name Note 99%
CAS 461-72-3
Chemical Name or Material Hydantoin
Molecular Formula C3H4N2O2
MDL Number MFCD00005259
Assay Percent Range 99%