missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Hydantoin-5-acetic acid, 98%

Manufacturer:  Alfa Aesar™ A1173106

 View more versions of this product

Catalog No. AAA1173106


 

Specifications

Specifications

Hydantoin-5-acetic acid
5g
98%
MFCD00005267
2,4-Imidazolidinedione-5-acetic acid
C(C1C(=O)NC(=O)N1)C(=O)O
158.113
158.11
≈217°C (dec.)
5427-26-9
C5H6N2O4
83035
DQQLZADYSWBCOX-UHFFFAOYSA-N
2-(2,5-dioxoimidazolidin-4-yl)acetic acid
95492

Chemical Identifiers

5427-26-9
158.113
DQQLZADYSWBCOX-UHFFFAOYSA-N
95492
C5H6N2O4
MFCD00005267
2,4-Imidazolidinedione-5-acetic acid
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

SDS Documents