missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Cytiva HyClone™ Water, Molecular Biology Grade
For critical applications involving nucleotides such as RNA and DNA. HyClone™ Water, Molecular Biology Grade contains no DEPC or other carcinogenic chemicals.
$13.69 - $169.25
Chemical Identifiers
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
SH3053801
|
Global Ls Soln Canada Ulc
SH3053801 |
100mL |
Each for $13.69
|
|
SH3053803
|
Global Ls Soln Canada Ulc
SH3053803 |
1000mL |
Each for $29.60
|
|
SH30538FS
|
Global Ls Soln Canada Ulc
SH30538FS |
6 x 500mL |
Pack of 6 Each for $106.18
|
|
SH3053802
|
Global Ls Soln Canada Ulc
SH3053802 |
500mL |
Each for $19.29
|
|
SH30538LS
|
Global Ls Soln Canada Ulc
SH30538LS |
6 x 1L |
Pack of 6 Each for $169.25
|
|
Description
- Format: Liquid
Chemical Identifiers
0.0 | |
icon-pdf.png |
Specifications
Manufactured in cGMP (21 CFR 820) compliant and ISO9001 certified facilities | |
5.0 to 7.0 | |
100mL |
Liquid | |
0.1μm sterile filtered |
Safety and Handling
Recommended Storage : 2° to 30°C