Please login to your online account to display your discounted pricing

HyClone™ Water, Molecular Biology Grade


For critical applications involving nucleotides such as RNA and DNA. HyClone™ Water, Molecular Biology Grade contains no DEPC or other carcinogenic chemicals.

Manufacturer: ge healthcare bio-science  SH30538.LS

 View more versions of this product

Catalog No. SH30538LS

  • / Pack of 6 Each
 

Description & Specifications

Specifications

Description Molecular Biology Grade Water
pH 5.0 to 7.0
Quantity 6 X 1000mL
State at Room Temperature Liquid
Sterilization Method Filter Sterilized - .1μm
Storage Requirements 2° to 30°C
Without Additives Dnase, Rnase, Protease
Certifications/Compliance Manufactured under cGMP and ISO 9001 certified facilities
Endotoxin Concentration 0.05EU/mL

  • Format: Liquid