Please login to your online account to display your discounted pricing

HyClone™ Water for Injection (WFI) Quality Water


Supports cell culture growth and biomanufacturing process

Manufacturer: ge healthcare bio-science  SH3022125

 View more versions of this product

Catalog No. SH3022125

  • / Each
 

Description & Specifications

Specifications

Physical State Liquid
Quantity 20L
Purity Grade WFI Grade
Purity Grade Notes Manufactured under cGMP and ISO 9001 certified facilities

  • Format: Liquid