Please login to your online account to display your discounted pricing

HyClone™ Trypsin Protease


Derived from porcine pancreas. HyClone™ Trypsin Protease is used to disaggregate adherent cells from substrate or surface to which they are attached.

Manufacturer: ge healthcare bio-science  SH3004201

 View more versions of this product

Catalog No. SH3004201

  • / Each
 

Description & Specifications

Specifications

Source Porcine pancreas
Sterilization Method 0.1μm sterile filtered
Shelf Life 24 months
Storage Requirements Store frozen.
With Additives Porcine trypsin, HBSS, EDTA
Concentration 0.25% (1X)
Format Liquid
Quantity 100mL
Without Additives Calcium and Magnesium
Description Trypsin, 0.25% 1X, with 2.5g porcine trypsin (1:250/L gamma irradiated) in HBSS with 0.2g/L EDTA, without calcium, magnesium (liquid)

  • Derived from porcine pancreas
  • Gamma irradiated prior to hydration and filling
  • Formulated without calcium and magnesium