Please login to your online account to display your discounted pricing

HyClone™ SFM4MegaVir™ Media


Manufacturer: ge healthcare bio-science  SH30552.02

 View more versions of this product

Catalog No. SH3055202

  • / Each
 

Description & Specifications

Specifications

Quantity 1000mL
Storage Requirements 2° to 8°C
Without Additives Protein, (L- Glutamine), Animal derived component free
Description SFM4MegaVir without L-Glutamine (Liquid)
Format Liquid

Increase process yields in the manufacture of viral vaccines in a variety of cell lines with HyClone™ SFM4MegaVir Cell Culture Media.