Please login to your online account to display your discounted pricing

HyClone™ SFM4HEK293™ Media


Manufacturer: ge healthcare bio-science  SH3052101

 View more versions of this product

Catalog No. SH3052101

  • / Each
 

Description & Specifications

Specifications

Description SFM4HEK293 Media
Quantity 500mL
Storage Requirements 2° to 8°C
With Additives L-Glutamine
Certifications/Compliance cGMP
Format Liquid

Support production of adenoviral vectors and recombinant proteins in HEK 239 cultures with HyClone™ SFM4HEK293™ Media, a versatile, protein-free cell culture media containing no animal-derived components.