HyClone™ SFM4CHO-Utility™ Media (Liquid) with L-glutamine
$65.16 - $65.16
Products ${productFamilyLength}
Catalog Number Mfr. No. Description Quantity Price Quantity  
Catalog Number Mfr. No. Description Quantity Price Quantity  
SH3051601 Ge Healthcare Bio-Science
SH3051601
SFM4CHO Liquid 500mL Each for $65.16
Specifications
Description
Support cell culture growth with HyClone™ SFM4CHO-Utility™ Media, a serum- and protein-free high performance media designed for superior growth of many CHO cells with minimal adaptation.