Please login to your online account to display your discounted pricing

HyClone™ SFM4CHO-Utility™ Media (Liquid) with L-glutamine


Manufacturer: ge healthcare bio-science  SH3051601

 View more versions of this product

Catalog No. SH3051601

  • / Each
 

Description & Specifications

Specifications

Quantity 500mL
Format Liquid
With Additives L-glutamine
Description SFM4CHO Liquid
Certifications/Compliance cGMP
Storage Requirements 2° to 8°C

Support cell culture growth with HyClone™ SFM4CHO-Utility™ Media, a serum- and protein-free high performance media designed for superior growth of many CHO cells with minimal adaptation.