Please login to your online account to display your discounted pricing

HyClone™ Phosphate Buffered Saline (PBS)


Buffer cell culture growth with PBS

Manufacturer: ge healthcare bio-science  SH3025602

 View more versions of this product

Catalog No. SH3025602

  • / Each
 

Description & Specifications

Specifications

Product Type 0.067M PO4 Phosphate Buffered Saline (PBS)
Format Liquid
Sterilization Method Filter Sterilized - .1μm
pH 7.0 to 7.2
Quantity 1000mL
Storage Requirements 15° to 3°C
Without Additives Calcium, Magnesium, Phenol Red
Certifications/Compliance cGMP and ISO 9001:2000 Guidelines
Concentration 1X
Endotoxin Concentration 1.0EU/mL

  • QC-tested solution