missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Cytiva (Formerly GE Healthcare Life Sciences) HyClone™ Newborn Calf Serum (U.S.)
Augment cell culture growth with U.S.-sourced HyClone™ Newborn Bovine Calf Serum.
$210.29 - $210.29
Specifications
Description | Newborn Calf Serum (U.S.) |
---|---|
Sterilization Method | Filtered through three sequential 100nm pore-size rated filters. |
Sterility | Sterile |
Catalog Number | Mfr. No. | Quantity | Processing | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Processing | Price | Quantity | ||||
SH3011803
|
Cytiva (Formerly GE Healthcare Life Sciences)
SH3011803 |
500mL |
Each for $210.29
|
|
Description
- Filtered through sequential 100nm filters and sourced from animals that are typically less than 10 days old
Specifications
Newborn Calf Serum (U.S.) | |
Filtered through three sequential 100nm pore-size rated filters. | |
Sterile |