Please login to your online account to display your discounted pricing
HyClone™ HyCell-STEM™ Media
Complete media for culturing human embryonic stem cells (hESCs) and induced pluripotent stem (hiPS) cells.
$138.73 - $138.73
Specifications
For Use With (Application) Culturing human embryonic stem cells (hESCs) and induced pluripotent stem (hiPS) cells
View More Specs
Includes:  HyCell-STEM kit includes 6x supplement and DMEM/F12 to create a complete media. Basic fibroblast growth factor must be added to the completed media at time of use.
Requires:  bFGF (10ng/ml) must be added to the media at time of use.
 Alert
Hyclone Cell Culture products now a part of GE Healthcare.
Products
Catalog Number Mfr. No. Media Type Quantity Price Quantity    
SR3000301
ge healthcare bio-science
SR3000301
HyCell-STEM 6x Supplement 100mL Each for $138.73
Description & Specifications
Specifications
For Use With (Application) Culturing human embryonic stem cells (hESCs) and induced pluripotent stem (hiPS) cells
HyCell-STEM Media is designed to help generate large numbers of human embryonic and induced pluripotent stem cells with fewer passages and is intended for use with stem cells on feeder layer cultures. HyCell-STEM is provided as a 6x supplement that is added to DMEM/F12 to create a complete media. Basic fibroblast growth factor is added to the completed media at time of use.
Highlights:
  • Increase rate of expansion
  • Sustain healthy stem cell morphology
  • Maintain stemness
  • Preserve pluripotency
  • Improved post-thaw recovery
HyCell-STEM Media is intended for use with stem cells on feeder layer cultures.