Please login to your online account to display your discounted pricing
HyClone™ HyCell-STEM™ Media
Manufacturer: ge healthcare bio-science  SR3000401
 View more versions of this product
Catalog No. SR3000401
  • / Each
Description & Specifications
Specifications
Description HyCell-STEM-FF 6x supplement
For Use With (Application) Culturing human embryonic stem cells (hESCs) and induced pluripotent stem (hiPS) cells
Product Type HyCell-STEM-FF 6x Supplement
Quantity 500mL
Media Type HyCell-STEM-FF 6x Supplement
Quantity 500mL
Culture human embryonic stem cells (hESCs) and induced pluripotent stem (hiPS) cells with HyCell-STEM™ Media, a complete media intended for use with stem cells on feeder layer cultures. This defined, serum-free formulation is used for expanding and maintaining stem cell colonies on feeder layer cultures with daily or every other day feeds, resulting in increased cell growth with fewer passages, all while maintaining pluripotency.