HyClone™ HyCell-STEM™ Media
$300.47 - $300.47
Products ${productFamilyLength}
Catalog Number Mfr. No. Description Price Quantity  
Catalog Number Mfr. No. Description Price Quantity  
SR3000401 Ge Healthcare Bio-Science
SR3000401
HyCell-STEM-FF 6x supplement Each for $300.47
Specifications
Description
Culture human embryonic stem cells (hESCs) and induced pluripotent stem (hiPS) cells with HyCell-STEM™ Media, a complete media intended for use with stem cells on feeder layer cultures. This defined, serum-free formulation is used for expanding and maintaining stem cell colonies on feeder layer cultures with daily or every other day feeds, resulting in increased cell growth with fewer passages, all while maintaining pluripotency.