Please login to your online account to display your discounted pricing

HyClone™ HyCell-STEM™ Media


Manufacturer: ge healthcare bio-science  SR30004KT

 View more versions of this product

Catalog No. SR30004KT

  • / Each
 

Description & Specifications

Specifications

Description HyCell-STEM-FF media kit
Quantity 600mL kit
Kit Contents 100mL 6x supplement and 500mL DMEM/F12
For Use With (Application) Culturing human embryonic stem cells (hESCs) and induced pluripotent stem (hiPS) cells
Product Type HyCell-STEM-FF Media Kit
Media Type HyCell-STEM-FF Media Kit
Quantity 600mL Kit

Culture human embryonic stem cells (hESCs) and induced pluripotent stem (hiPS) cells with HyCell-STEM™ Media, a complete media intended for use with stem cells on feeder layer cultures. This defined, serum-free formulation is used for expanding and maintaining stem cell colonies on feeder layer cultures with daily or every other day feeds, resulting in increased cell growth with fewer passages, all while maintaining pluripotency.