Please login to your online account to display your discounted pricing

HyClone™ G418 Sulfate


Manufacturer: ge healthcare bio-science  SV30069.01

 View more versions of this product

Catalog No. SV3006901

  • / Each
 

Description & Specifications

Specifications

Antibiotic Spectrum Selective agent for eukaryotic cells
Concentration 50mg/mL
Description G418 Sulfate Solution, 50mg/mL
Format Liquid
Purity >9%
Quantity 20mL (50mg/mL)

Control or destroy destructive microorganisms with HyClone™ G418 Sulfate.