Please login to your online account to display your discounted pricing

HyClone™ G418 Sulfate

Manufacturer: ge healthcare bio-science  SV30068.02

 View all options for this product

Catalog No. SV3006802


Description & Specifications


Antibiotic Spectrum Selective agent for eukaryotic cells
Concentration 5mg/mL
Description G418 Sulfate, >90% Purity
Format Powder
Purity >90%
Quantity 5g
Storage 2° to 8°C

Control or destroy destructive microorganisms with HyClone™ G418 Sulfate.