Please login to your online account to display your discounted pricing
Fetal Bovine Serum (U.S.), Super Low IgG
Processed using protein G chromatography to reduce the levels to those required for monoclonal antibody production
Manufacturer: ge healthcare bio-science  SH3089803IH
 View more versions of this product
 Alert
Hyclone Cell Culture products now a part of GE Healthcare.
Catalog No. SH3089803IH
  • / Each
Description & Specifications
Specifications
Heat Inactivated Yes
Storage 2-8°C
Quantity 500mL