missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Cytiva (Formerly GE Healthcare Life Sciences) HyClone™ Fetal Bovine Serum (U.S.), Defined
Used as serum-supplement for the in vitro cell culture of eukaryotic cells
$436.04 - $1,862.06
Specifications
Certifications/Compliance | Manufactured in cGMP (21 CFR 820) compliant and ISO9001 certified facilities |
---|---|
Endotoxin Concentration | ≤ 10 EU/mL |
Grade | Defined |
Hemoglobin Concentration | ≤ 10 mg/dL |
Origin | U.S. |
Catalog Number | Mfr. No. | Quantity | Processing | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Processing | Price | Quantity | ||||
SH3007002
|
Cytiva (Formerly GE Healthcare Life Sciences)
SH3007002 |
100mL | None |
Each for $436.04
|
|
SH3007003HI
|
Cytiva (Formerly GE Healthcare Life Sciences)
SH3007003HI |
500mL | Heat Inactivated |
Each for $1,862.06
|
|
SH3007003
|
Cytiva (Formerly GE Healthcare Life Sciences)
SH3007003 |
500mL | None |
Each for $1,780.76
|
|
Description
- Maximize cell culture growth
- Tested for endotoxin, hemoglobin, and additional biochemical assays
- Filtered through serial 40nm pore-size rated filters
Specifications
Manufactured in cGMP (21 CFR 820) compliant and ISO9001 certified facilities | |
Defined | |
U.S. | |
-10°C | |
USDA Inspected |
≤ 10 EU/mL | |
≤ 10 mg/dL | |
No | |
Sterile |