Please login to your online account to display your discounted pricing

HyClone™ Fetal Bovine Serum (U.S.), Characterized


Used as serum-supplement for the in vitro cell culture of eukaryotic cells

Manufacturer: ge healthcare bio-science  SH3007103IR

 View more versions of this product

Catalog No. SH3007103IR

  • / Each
 

Description & Specifications

Specifications

Manufacturing Origin U.S.
Tested For Bluetongue
Tested For Bovine Adenovirus
Tested For Bovine Parvovirus
Tested For Bovine RSV
Tested For BVDV
Tested For Cytopathogenic Agents
Tested For Hemadsorbing Agents
Tested For Rabies Virus
Tested For Reovirus
Additional Assays Albumin
Mycoplasma Detected No
Certifications/Compliance Manufactured under cGMP and ISO 9001 certified facilities.
USDA Safety Testing Included USDA Inspected
Processing Irradiated
Sterilization Method Filtered through three sequential 100nm pore-size rated filters; Irradiated
Storage -10°C
Heat Inactivated No
Sterile Yes
Description Characterized Fetal Bovine Serum, US Origin
Endotoxin Concentration ≤25EU/mL
Quantity 500mL
Hemoglobin Concentration ≤25mg/dL
Grade Characterized

  • Enhance growth of cells
  • Tested for endotoxin, hemoglobin, and additional biochemical assays
  • Filtered through serial 100nm pore-size rated filters