Please login to your online account to display your discounted pricing

HyClone™ Dulbecco's Phosphate Buffered Saline, Powder


Manufacturer: ge healthcare bio-science  SH3001303

 View more versions of this product

Catalog No. SH3001303

  • / Each
 

Description & Specifications

Specifications

Storage Requirements 2° to 8°C
Certifications/Compliance Manufactured under cGMP and ISO 9001 certified facilities
Without Additives Calcium, Magnesium
Quantity 1 X 10L
State at Room Temperature Solid
Description Without Calcium, Magnesium (Powder), 10L

Buffer cell culture with HyClone™ Dulbecco's Phosphate Buffered Saline (PBS), Powder.