Please login to your online account to display your discounted pricing

HyClone™ Dulbecco's Modified Eagles Medium


Designed to support cell culture growth and biomanufacturing processes.

Manufacturer: ge healthcare bio-science  SH30262.01

 View more versions of this product

Catalog No. SH3026201

  • / Each
 

Description & Specifications

Specifications

Format Liquid
Product Type DMEM media
Shelf Life 18 months
With Additives L-glucose
Without Additives L-glutamine, sodium pyruvate, Calcium, Magnesium
Storage 2° to 8°C, away from light

  • Format: Liquid

Accessories

18 months