Cytiva HyClone™ Calf Serum (New Zealand)
Manufacturer:  Cytiva SH3040101
 View more versions of this product
Catalog No. SH3040101
Description
Description
Supplement cell culture growth with New Zealand-sourced HyClone™ Bovine Calf Serum, an excellent and cost-effective alternative to fetal bovine serum (FBS).
Specifications
Specifications
Albumin
Sera should be stored at -10°C or lower. Once thawed, sera should be stored at 2° to 8°C.
500 mL
Bluetongue,Bovine Adenovirus,Bovine Parvovirus,Bovine RSV,BVDV,Cytopathogenic Agents,Hemadsorbing Agents,Rabies Virus,Reovirus
Yes
<-1°C
USDA Regulations
Calf Serum (New Zealand)
Sterile
≤ 25 EU/mL
Sera should be stored at -10°C or lower. Once thawed, sera should be stored at 2° to 8°C.
Yes