missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Boekel Scientific™ Hybridization Glass Bottles for Boekel Little Shot Oven
Manufacturer: Boekel Scientific™ 5020225
Specifications
225 mm | |
Hybridization Bottle | |
1.37 in. | |
Boekel little SHOT III medium capacity oven | |
8.85 in. |
Glass | |
Blue | |
35 mm | |
Blue caps and silicone O-ring |