missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Boekel Scientific™ Hybridization Glass Bottles for Boekel Little Shot Oven

Manufacturer:  Boekel Scientific™ 5020150

 View more versions of this product

Catalog No. 11-700-111



Description

Description

Specifications

Specifications

150 mm
Hybridization Bottle
1.37 in.
Boekel little SHOT III medium capacity oven
5.9 in.
Glass
Blue
35 mm
Blue caps and silicone O-ring
SDS
Documents
Product Certifications
Promotions

Promotions