Please login to your online account to display your discounted pricing

Alfa Aesar™ Hyaluronic acid sodium salt, Streptococcus equi, 91%


Quantity 1g

Manufacturer: alfa aesar™  J6699303

 View more versions of this product

Catalog No. AAJ6699303

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Chemical Name or Material Hyaluronic acid sodium salt
CAS 9067-32-7
Physical Form Powder
MDL Number MFCD01773053
Assay 91%
Name Note Streptococcus equi
Merck Index 14,4757
Solubility Information Soluble in water
Quantity 1g
RTECS Number MT7250000
Storage Note 1 -30° to -10°C
Color White

Specifications

Chemical Name or Material Hyaluronic acid sodium salt
CAS 9067-32-7
Name Note Streptococcus equi
MDL Number MFCD01773053
Merck Index 14,4757

Applications
Bacterial hyaluronic acid (HA) is a natural non-sulfated high molecular weight glycosaminoglycan that may be used to study formulations of non-animal gels potentially useful for procedures such as soft tissue augmentation.

Solubility
Soluble in water

Notes
Research Use only: Not intended for animal or human diagnostic or therapeutic use. Molecular weight is greater than or equal to 1MDa

Accessories

-30° to -10°C

SDS & Safety Documents