missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Hyaluronic acid, bovine vitreous humor

A natural high-viscosity polymer of acetylglucosamine and glucuronic acid

$212.81 - $1,489.69

Chemical Identifiers

CAS 9004-61-9
Molecular Formula C28H44N2O23
Molecular Weight (g/mol) 776.651
MDL Number MFCD00131348
InChI Key KIUKXJAPPMFGSW-YXBJCWEESA-N
Synonym hyaluronic acid, dsstox_cid_26750, dsstox_rid_81875, dsstox_gsid_46750, 4xi-alpha-d-xylo-hexopyranuronosyl-1->3-3xi-2-acetylamino-2-deoxy-beta-d-ribo-hexopyranosyl-1->4-5xi-beta-d-xylo-hexopyranuronosyl-1->3-2-acetylamino-2-deoxy-d-glucopyranose
PubChem CID 24728612
IUPAC Name (2S,4S,5R,6S)-6-[(2S,3R,5S,6R)-3-acetamido-2-[(3S,4R,5R,6R)-6-[(3R,4R,5S,6R)-3-acetamido-2,5-dihydroxy-6-(hydroxymethyl)oxan-4-yl]oxy-2-carboxy-4,5-dihydroxyoxan-3-yl]oxy-5-hydroxy-6-(hydroxymethyl)oxan-4-yl]oxy-3,4,5-trihydroxyoxane-2-carboxylic acid
SMILES CC(=O)NC1C(C(C(OC1O)CO)O)OC2C(C(C(C(O2)C(=O)O)OC3C(C(C(C(O3)CO)O)OC4C(C(C(C(O4)C(=O)O)O)O)O)NC(=O)C)O)O
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAJ60566MA View Documents Alfa Aesar™
J60566MA
10mg Each for $212.81
 
AAJ60566MF View Documents Alfa Aesar™
J60566MF
50mg Each for $766.43
 
AAJ60566MC View Documents Alfa Aesar™
J60566MC
100mg Each for $1,489.69
 
Description

Description

Applications
Hyaluronic acid is a naturally derived, non - immunogenic, non - adhesive glycosaminoglycan that plays a prominent role in various wound - healing processes, as it as it is naturally angiogenic when degraded to small fragments. Hyaluronic acid promotes early inflammation which is critical for initiating wound healing, but then moderates later stages of the process, allowing matrix stabilization and reduction of long term inflammation. Hyaluronic acid is a main source for pharmaceutical, medical and cosmetic application.

Solubility
Soluble in water.

Notes
Supplied as a partially purified powder, suitable as a substrate for hyaluronidase
Specifications

Chemical Identifiers

9004-61-9
776.651
KIUKXJAPPMFGSW-YXBJCWEESA-N
24728612
CC(=O)NC1C(C(C(OC1O)CO)O)OC2C(C(C(C(O2)C(=O)O)OC3C(C(C(C(O3)CO)O)OC4C(C(C(C(O4)C(=O)O)O)O)O)NC(=O)C)O)O
C28H44N2O23
MFCD00131348
hyaluronic acid, dsstox_cid_26750, dsstox_rid_81875, dsstox_gsid_46750, 4xi-alpha-d-xylo-hexopyranuronosyl-1->3-3xi-2-acetylamino-2-deoxy-beta-d-ribo-hexopyranosyl-1->4-5xi-beta-d-xylo-hexopyranuronosyl-1->3-2-acetylamino-2-deoxy-d-glucopyranose
(2S,4S,5R,6S)-6-[(2S,3R,5S,6R)-3-acetamido-2-[(3S,4R,5R,6R)-6-[(3R,4R,5S,6R)-3-acetamido-2,5-dihydroxy-6-(hydroxymethyl)oxan-4-yl]oxy-2-carboxy-4,5-dihydroxyoxan-3-yl]oxy-5-hydroxy-6-(hydroxymethyl)oxan-4-yl]oxy-3,4,5-trihydroxyoxane-2-carboxylic acid

Specifications

White
9004-61-9
MFCD00131348
hyaluronic acid, dsstox_cid_26750, dsstox_rid_81875, dsstox_gsid_46750, 4xi-alpha-d-xylo-hexopyranuronosyl-1->3-3xi-2-acetylamino-2-deoxy-beta-d-ribo-hexopyranosyl-1->4-5xi-beta-d-xylo-hexopyranuronosyl-1->3-2-acetylamino-2-deoxy-d-glucopyranose
CC(=O)NC1C(C(C(OC1O)CO)O)OC2C(C(C(C(O2)C(=O)O)OC3C(C(C(C(O3)CO)O)OC4C(C(C(C(O4)C(=O)O)O)O)O)NC(=O)C)O)O
776.651
Lyophilized Powder
10mg
C28H44N2O23
14,4757
KIUKXJAPPMFGSW-YXBJCWEESA-N
(2S,4S,5R,6S)-6-[(2S,3R,5S,6R)-3-acetamido-2-[(3S,4R,5R,6R)-6-[(3R,4R,5S,6R)-3-acetamido-2,5-dihydroxy-6-(hydroxymethyl)oxan-4-yl]oxy-2-carboxy-4,5-dihydroxyoxan-3-yl]oxy-5-hydroxy-6-(hydroxymethyl)oxan-4-yl]oxy-3,4,5-trihydroxyoxane-2-carboxylic acid
24728612
Hyaluronic acid, bovine vitreous humor
Safety and Handling

Safety and Handling

TSCA : TSCA

Recommended Storage : 2° to 8°C