missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Humboldt Meker-Type Burner

Provides extremely hot flame

Manufacturer:  Humboldt H5600*

Catalog No. 22-043-331



Description

Description

High-temperature Meker-type burners are for use where an extremely hot flame is required. The Meker cap forms many short, small, uniform blue cones (no cold cones). Uses LP gas.

  • Specifically designed to prevent back-flashing—an important safety feature
  • One-piece nickel-plated Venturi tube and an octagonal base of heavy, die-cast nickel-plated zinc alloy
  • Base measures 3.5 in. (8.75cm) in diameter
  • Capable of heat output in excess of 12,000 BTU per hour
  • Wing top is not available for this burner

Specifications

Specifications

38 mm
8.5 in.
LPG
10,980 BTU
216 mm
High Temperature Meker Burner
SDS
Documents