Humboldt Filter Pump Aspirators
Offer excellent performance for vacuum filtrations under both high and low water pressure conditions
Supplier: Humboldt H12020
Description
- Swirling type, available in two models: Chapman and Short
- Nickel-plated
- Ball-type, positive-action valve in suction tube prevents water backflow if pressure is reduced
- Water inlet: 3/8 in., NPT tapered
Specifications
Nickel-plated Zinc | |
5.38 in. | |
137 mm |