missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human ZNF84 (aa 46-188) Control Fragment Recombinant Protein

Catalog No. RP99736
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
RP99736 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. RP99736 Supplier Invitrogen™ Supplier No. RP99736
Only null left

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (31%), Rat (31%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-55992 (PA5-55992. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

ZNF84, a gene located on chromosome 12, encodes a zinc finger protein whose function is undescribed.

Specifications

Accession Number P51523
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 7637
Name Human ZNF84 (aa 46-188) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias HPF2; zinc finger protein 84; Zinc finger protein HPF2; ZNF84
Common Name ZNF84
Gene Symbol ZNF84
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence SLGYEVMKPDVIFKLEQGEEPWVGDGEIPSSDSPEVWKVDGNMMWHQDNQDKLKIIKRGHECDAFGKNFNLNMNFVPLRKSNSEGDLDGLILKHHLDLLIPKGDYGKAESDDFNVFDNFFLHSKPEDTDTWLKYYDCDKYKES
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less